Product Details
- SNP ID
-
rs3897842
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:213058153 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTATTCGCAGGTGTGATCATAGTG[G/T]GCTACAGCCTCGACCTCCTAGGCTG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RPS6KC1
PubMed Links
Gene Details
- Gene
- RPS6KC1
- Gene Name
- ribosomal protein S6 kinase C1
There are no transcripts associated with this gene.
View Full Product Details