Product Details

SNP ID
rs6681851
Assay Type
Functionally tested
NCBI dbSNP Submissions
42
Location
Chr.1:212829526 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTCCTTATTCAAATAATCAAAAC[C/T]ATAACTTTAACAGAGGAAGTTAATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA45 PubMed Links

Gene Details

Gene
SPATA45
Gene Name
spermatogenesis associated 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024601.2 Intron NP_001019772.1

View Full Product Details