Product Details
- SNP ID
-
rs35810960
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:121755872 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGAGCCCCCTCCCTTCCCCATTCCC[C/T]TTGCCTTTCCATGCCTAGTTGGGCC
- Phenotype
-
MIM: 600575
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSN
PubMed Links
Gene Details
- Gene
- TSN
- Gene Name
- translin
View Full Product Details