Product Details
- SNP ID
-
rs187192
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:88194409 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAATCAACCACTGATGGCCACTTA[G/T]GTTGACCCCATGACTTTGCCACTGT
- Phenotype
-
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM161B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75762615] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM161B
- Gene Name
- transmembrane protein 161B
View Full Product Details