Product Details

SNP ID
rs11107041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93468886 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGAATACTCTGTGTACCATTTGT[C/G]AGTGACTGTTTCAAGAGTAATTACA
Phenotype
MIM: 611847
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL42 PubMed Links

Gene Details

Gene
MRPL42
Gene Name
mitochondrial ribosomal protein L42
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014050.3 Intron NP_054769.1
NM_172177.3 Intron NP_751917.1

View Full Product Details