Product Details

SNP ID
rs11757034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32038945 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCACCTCAGCCTCAAGTGTGAGCCA[C/T]CTTTGGGGCATCCCCAATCCAGGTC
Phenotype
MIM: 120820 MIM: 613815 MIM: 600985
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C4B PubMed Links

Gene Details

Gene
C4B
Gene Name
complement component 4B (Chido blood group)
There are no transcripts associated with this gene.

Gene
CYP21A2
Gene Name
cytochrome P450 family 21 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000500.7 Intron NP_000491.4
NM_001128590.3 Intron NP_001122062.3
Gene
TNXB
Gene Name
tenascin XB
There are no transcripts associated with this gene.

View Full Product Details