Product Details
- SNP ID
-
hCV27305426
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:46856403 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTAATGCCTTGAATCCCCCCAGAAG[A/T]TTGCTTCTCATTGAACGTTTGATTT
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRF5
PubMed Links
Gene Details
- Gene
- ADGRF5
- Gene Name
- adhesion G protein-coupled receptor F5
View Full Product Details