Product Details

SNP ID
rs910056416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:16671825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACAATTCTGAGACAGGCTGAA[A/G]CAAACAAGAAAAAACCATATGTATT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC144A PubMed Links

Gene Details

Gene
CCDC144A
Gene Name
coiled-coil domain containing 144A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014695.2 Intron NP_055510.1

View Full Product Details