Product Details

SNP ID
rs4423879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:104476417 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAGTCTAATAAAACATCAGCTC[C/T]TAAAAGCTGAAAATGGGGCATCTCT
Phenotype
MIM: 611645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CXXC4 PubMed Links

Gene Details

Gene
CXXC4
Gene Name
CXXC finger protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025212.3 Intron NP_079488.2

View Full Product Details