Product Details

SNP ID
rs3013640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:29512790 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGAGCAGGCCTGATACGGAGGC[C/T]CAGCCCATGGCCATCCACCCGCCCT
Phenotype
MIM: 104615
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MTUS2 PubMed Links

Gene Details

Gene
MTUS2
Gene Name
microtubule associated tumor suppressor candidate 2
There are no transcripts associated with this gene.

Gene
SLC7A1
Gene Name
solute carrier family 7 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003045.4 3978 UTR 3 NP_003036.1
XM_005266507.3 3978 UTR 3 XP_005266564.1
XM_017020713.1 3978 UTR 3 XP_016876202.1
XM_017020714.1 3978 UTR 3 XP_016876203.1
XM_017020715.1 3978 UTR 3 XP_016876204.1
XM_017020716.1 3978 UTR 3 XP_016876205.1

View Full Product Details