Product Details

SNP ID
rs14072
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:138090801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAACAAGTTTAGAATACCTGGT[A/T]TATTGGGAAAACTTCATAATGAAAA
Phenotype
MIM: 609301
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PERP PubMed Links

Gene Details

Gene
PERP
Gene Name
PERP, TP53 apoptosis effector
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022121.4 2006 UTR 3 NP_071404.2

View Full Product Details