Product Details

SNP ID
rs7489608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:114323191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTAGAACATTGTAGGACACAAAGT[A/T]ATTTATTTATTAAATGAATTAATAG
Phenotype
MIM: 616327
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CHAMP1 PubMed Links

Gene Details

Gene
CHAMP1
Gene Name
chromosome alignment maintaining phosphoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164144.1 Intron NP_001157616.1
NM_001164145.1 Intron NP_001157617.1
NM_032436.2 Intron NP_115812.1
Gene
LINC01054
Gene Name
long intergenic non-protein coding RNA 1054
There are no transcripts associated with this gene.

View Full Product Details