Product Details

SNP ID
rs2831348
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:28015959 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTATCTAACACTCCCCCTTTCTCCT[A/C]AAGTTTTGGTCAAGGTTTTATTTGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC00314 PubMed Links

Gene Details

Gene
LINC00314
Gene Name
long intergenic non-protein coding RNA 314
There are no transcripts associated with this gene.

View Full Product Details