Product Details

SNP ID
rs13734
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:17614084 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCTCGGAGCTACCGGAAGTTGG[A/G]CCTGGATAACGCTGTGTAGGTTGGT
Phenotype
MIM: 609114 MIM: 601418
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DSTN PubMed Links

Gene Details

Gene
DSTN
Gene Name
destrin, actin depolymerizing factor
There are no transcripts associated with this gene.

Gene
RRBP1
Gene Name
ribosome binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042576.1 3386 UTR 3 NP_001036041.1
NM_004587.2 3386 UTR 3 NP_004578.2

View Full Product Details