Product Details

SNP ID
rs10485582
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:18589047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATATGCGTTCTTGTGTCCACAAA[A/G]GTATGAATTAAAAACAAAACAGGCC
Phenotype
MIM: 610996
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DTD1 PubMed Links

Gene Details

Gene
DTD1
Gene Name
D-tyrosyl-tRNA deacylase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318043.1 Intron NP_001304972.1
NM_080820.5 Intron NP_543010.3
XM_005260872.4 Intron XP_005260929.1

View Full Product Details