Product Details

SNP ID
rs6042390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:13854377 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATTTCTTCCATGTAAAAAGGTT[G/T]TCATATGTTTCTCGTACTCCATGGT
Phenotype
MIM: 614289
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEL1L2 PubMed Links

Gene Details

Gene
SEL1L2
Gene Name
SEL1L2 ERAD E3 ligase adaptor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271539.1 Intron NP_001258468.1
XM_006723646.2 Intron XP_006723709.1
XM_006723647.2 Intron XP_006723710.1
XM_006723648.2 Intron XP_006723711.1
XM_006723650.2 Intron XP_006723713.1
XM_006723651.2 Intron XP_006723714.1
XM_006723652.2 Intron XP_006723715.1
XM_006723654.3 Intron XP_006723717.1
XM_006723655.2 Intron XP_006723718.1
XM_006723656.2 Intron XP_006723719.1
XM_011529374.1 Intron XP_011527676.1
XM_011529375.1 Intron XP_011527677.1
XM_011529376.1 Intron XP_011527678.1
XM_011529377.1 Intron XP_011527679.1
XM_011529378.1 Intron XP_011527680.1
XM_011529379.1 Intron XP_011527681.1
XM_017028084.1 Intron XP_016883573.1

View Full Product Details