Product Details

SNP ID
rs2679203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:156437055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGAATGAAGTGCTGCTCTTAGCA[A/G]ATGGTCTCTAGTCTGGCTTCCACCT
Phenotype
MIM: 138430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPD2 PubMed Links

Gene Details

Gene
GPD2
Gene Name
glycerol-3-phosphate dehydrogenase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000408.4 Intron NP_000399.3
NM_001083112.2 Intron NP_001076581.2
XM_005246469.2 Intron XP_005246526.1
XM_011510977.2 Intron XP_011509279.1
XM_011510978.2 Intron XP_011509280.1
XM_011510980.2 Intron XP_011509282.1
XM_017003830.1 Intron XP_016859319.1
XM_017003831.1 Intron XP_016859320.1

View Full Product Details