Product Details

SNP ID
rs1135848
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:135095303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTTCAGGCAGCAGGGGTGAAAA[A/G]GCAGGGACAGGAGCAAATGCTGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM10L2B PubMed Links

Gene Details

Gene
SMIM10L2B
Gene Name
small integral membrane protein 10 like 2B
There are no transcripts associated with this gene.

View Full Product Details