Product Details

SNP ID
rs3847482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:117199308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACTCCAGCCTGGATGACAGAGCC[A/G]GAGATGTCTCAAAAATAAATAAGTA
Phenotype
MIM: 613655
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNK18 PubMed Links

Gene Details

Gene
KCNK18
Gene Name
potassium two pore domain channel subfamily K member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181840.1 Intron NP_862823.1

View Full Product Details