Product Details
- SNP ID
-
rs4516983
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:110250090 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCATCCACCCAGAACTGCAGAGATT[C/T]GAGGAAAGGGGAGGAAGAGGCATGG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
METTL24
PubMed Links
Gene Details
- Gene
- METTL24
- Gene Name
- methyltransferase like 24
View Full Product Details