Product Details

SNP ID
rs11970338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:98872474 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTACTTACAATCAACTTTAATGA[C/T]ACGAAACATTAACTTTGATCCTCCC
Phenotype
MIM: 605654
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXL4 PubMed Links

Gene Details

Gene
FBXL4
Gene Name
F-box and leucine rich repeat protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278716.1 4095 Intron NP_001265645.1
NM_012160.4 4095 Intron NP_036292.2
XM_005266930.2 4095 UTR 3 XP_005266987.1
XM_011535748.2 4095 Intron XP_011534050.1
XM_017010726.1 4095 UTR 3 XP_016866215.1
XM_017010727.1 4095 UTR 3 XP_016866216.1
XM_017010728.1 4095 UTR 3 XP_016866217.1

View Full Product Details