Product Details

SNP ID
rs3094070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30264052 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGCATCTGGGGAAAGTCATTGAA[C/G]TAAAATCAGAAAAATCTCTAGCCTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HCG17 PubMed Links

Gene Details

Gene
HCG17
Gene Name
HLA complex group 17 (non-protein coding)
There are no transcripts associated with this gene.

Gene
HLA-L
Gene Name
major histocompatibility complex, class I, L (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details