Product Details

SNP ID
rs3106065
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:131599597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAAAACATGAAACTAAAGAAATA[C/T]TTTACATGCAGATTCTTGCTTTATA
Phenotype
MIM: 611266
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POTEI PubMed Links

Gene Details

Gene
POTEI
Gene Name
POTE ankyrin domain family member I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277406.1 Intron NP_001264335.1
XM_011511670.2 Intron XP_011509972.1
XM_017004732.1 Intron XP_016860221.1
XM_017004733.1 Intron XP_016860222.1
XM_017004734.1 Intron XP_016860223.1
XM_017004735.1 Intron XP_016860224.1
Gene
POTEKP
Gene Name
POTE ankyrin domain family member K, pseudogene
There are no transcripts associated with this gene.

View Full Product Details