Product Details

SNP ID
rs3134021
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:93760440 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGATGCTGGGAAAACTGAGAAAA[C/T]ACCCAGGTTGGGTTTCTACCTTATT
Phenotype
MIM: 609884
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM67 PubMed Links

Gene Details

Gene
TMEM67
Gene Name
transmembrane protein 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142301.1 Intron NP_001135773.1
NM_153704.5 Intron NP_714915.3
XM_006716686.3 Intron XP_006716749.1
XM_011517363.2 Intron XP_011515665.1

View Full Product Details