Product Details

SNP ID
rs3124665
Assay Type
Functionally tested
NCBI dbSNP Submissions
66
Location
Chr.1:212935754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCGCTTTGCCTTATCTGAGTCTCA[A/G]CAGGCTTTTCCACGATATTATCAGG
Phenotype
MIM: 610471
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VASH2 PubMed Links

Gene Details

Gene
VASH2
Gene Name
vasohibin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136474.2 179 Intron NP_001129946.1
NM_001136475.2 179 Intron NP_001129947.1
NM_001301056.1 179 Intron NP_001287985.1
NM_024749.4 179 Intron NP_079025.2
XM_011509987.2 179 UTR 5 XP_011508289.1
XM_011509988.2 179 UTR 5 XP_011508290.1
XM_011509989.2 179 Intron XP_011508291.1
XM_011509991.2 179 Intron XP_011508293.2
XM_011509994.1 179 Intron XP_011508296.1
XM_011509995.1 179 Intron XP_011508297.1
XM_011509997.1 179 UTR 5 XP_011508299.1
XM_017002351.1 179 Intron XP_016857840.1
XM_017002352.1 179 Intron XP_016857841.1

View Full Product Details