Product Details

SNP ID
rs3177436
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1984945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGCTGAAGACCTAAGAAAAAGG[C/T]AAGATGTGTTTGCACGCAGCAGAGC
Phenotype
MIM: 600924 MIM: 611256 MIM: 603927 MIM: 605915
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFER PubMed Links

Gene Details

Gene
GFER
Gene Name
growth factor, augmenter of liver regeneration
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005262.2 Intron NP_005253.3
Gene
NOXO1
Gene Name
NADPH oxidase organizer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267721.1 Intron NP_001254650.1
NM_144603.3 Intron NP_653204.1
NM_172167.2 Intron NP_751907.1
NM_172168.2 Intron NP_751908.1
Gene
SYNGR3
Gene Name
synaptogyrin 3
There are no transcripts associated with this gene.

Gene
TBL3
Gene Name
transducin beta like 3
There are no transcripts associated with this gene.

View Full Product Details