Product Details

SNP ID
rs3178210
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12669562 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTACCCAGAGAACGGAGATTTAG[C/G]AGAAGCCAGAAGTCTTCCTTTCAAA
Phenotype
MIM: 600944 MIM: 609458 MIM: 616850
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DHPS PubMed Links

Gene Details

Gene
DHPS
Gene Name
deoxyhypusine synthase
There are no transcripts associated with this gene.

Gene
MAN2B1
Gene Name
mannosidase alpha class 2B member 1
There are no transcripts associated with this gene.

Gene
WDR83
Gene Name
WD repeat domain 83
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099737.2 90 Intron NP_001093207.1
NM_032332.3 90 Intron NP_115708.1
Gene
WDR83OS
Gene Name
WD repeat domain 83 opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016145.3 90 UTR 5 NP_057229.1
XM_017026864.1 90 UTR 5 XP_016882353.1

View Full Product Details