Product Details

SNP ID
rs3178259
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:152971174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTTAATAAGAGATCACAATCTTC[A/G]TAAATTCTTTTTGCTTTTCTTTGTA
Phenotype
MIM: 606013
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO5 PubMed Links

Gene Details

Gene
FBXO5
Gene Name
F-box protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142522.1 1442 Nonsense Mutation CGA,TGA R399* NP_001135994.1
NM_012177.3 1442 Nonsense Mutation CGA,TGA R445* NP_036309.1

View Full Product Details