Product Details
- SNP ID
-
rs1129208
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:18592314 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCCTGCAGTTCCAGACACCTCCC[A/C]CAATAAAGAGCTCCTCCTCTGTAGC
- Phenotype
-
MIM: 604237
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
C19orf60
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112466988] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C19orf60
- Gene Name
- chromosome 19 open reading frame 60
- Gene
- CRLF1
- Gene Name
- cytokine receptor like factor 1
View Full Product Details