Product Details
- SNP ID
-
rs3178587
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:156073827 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTTAATTCTGGATTCCCCCTCCT[C/T]ACTTGCCCCTGCGACAGGGCAGGGG
- Phenotype
-
MIM: 150330
MIM: 611007
MIM: 612942
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LMNA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78447160] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LMNA
- Gene Name
- lamin A/C
There are no transcripts associated with this gene.
- Gene
- MEX3A
- Gene Name
- mex-3 RNA binding family member A
- Gene
- RAB25
- Gene Name
- RAB25, member RAS oncogene family
There are no transcripts associated with this gene.
View Full Product Details