Product Details

SNP ID
rs3176453
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:50969978 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCCGATCTATAGTCCCTTGAGT[C/T]TCCACACACACTCCTCACAGAGATC
Phenotype
MIM: 603369 MIM: 604460
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKN2C PubMed Links

Gene Details

Gene
CDKN2C
Gene Name
cyclin dependent kinase inhibitor 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001262.2 9 Intron NP_001253.1
NM_078626.2 9 UTR 5 NP_523240.1
Gene
FAF1
Gene Name
Fas associated factor 1
There are no transcripts associated with this gene.

View Full Product Details