Product Details
- SNP ID
-
rs3176908
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:118886492 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGGCAGGGGCTGGGGGGTGGGGA[C/G]GGGCTGCTAATTCTCATCAATGAAA
- Phenotype
-
MIM: 609004
MIM: 601613
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BCL9L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs143478793] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCL9L
- Gene Name
- B-cell CLL/lymphoma 9-like
There are no transcripts associated with this gene.
- Gene
- CXCR5
- Gene Name
- C-X-C motif chemokine receptor 5
- Gene
- LOC101929133
- Gene Name
- uncharacterized LOC101929133
There are no transcripts associated with this gene.
View Full Product Details