Product Details

SNP ID
rs3180253
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103216425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTAGGGCAGATGATGGAAATCA[A/T]GAGAAAGACTAGGGAACAGCAGATG
Phenotype
MIM: 300692
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BEX4 PubMed Links

Gene Details

Gene
BEX4
Gene Name
brain expressed X-linked 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080425.3 424 Missense Mutation AAG,ATG K91M NP_001073894.1
NM_001127688.2 424 Missense Mutation AAG,ATG K91M NP_001121160.1

View Full Product Details