Product Details

SNP ID
rs3171692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41246208 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAAGGACAGCACGCGGGCAACTCC[C/T]TGGGTTCTGGCTCCGCCAGGGAGCC
Phenotype
MIM: 616236 MIM: 602362
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHADL PubMed Links

Gene Details

Gene
CHADL
Gene Name
chondroadherin like
There are no transcripts associated with this gene.

Gene
MIR6889
Gene Name
microRNA 6889
There are no transcripts associated with this gene.

Gene
RANGAP1
Gene Name
Ran GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278651.1 2591 UTR 3 NP_001265580.1
NM_001317930.1 2591 UTR 3 NP_001304859.1
NM_002883.3 2591 UTR 3 NP_002874.1
XM_005261695.1 2591 UTR 3 XP_005261752.1
XM_005261696.1 2591 UTR 3 XP_005261753.1
XM_006724289.3 2591 UTR 3 XP_006724352.1
XM_011530293.1 2591 UTR 3 XP_011528595.1
XM_011530294.2 2591 UTR 3 XP_011528596.1
XM_011530295.2 2591 UTR 3 XP_011528597.1
XM_011530297.1 2591 Intron XP_011528599.1
XM_017028893.1 2591 UTR 3 XP_016884382.1
XM_017028894.1 2591 UTR 3 XP_016884383.1
XM_017028895.1 2591 UTR 3 XP_016884384.1
XM_017028896.1 2591 UTR 3 XP_016884385.1
XM_017028897.1 2591 UTR 3 XP_016884386.1
XM_017028898.1 2591 UTR 3 XP_016884387.1
XM_017028899.1 2591 UTR 3 XP_016884388.1

View Full Product Details