Product Details

SNP ID
rs3180837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:150766337 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTAACACCATCAGAAGACAAGAAT[C/T]TGCTCGGTGCCATTTTATTTAATGC
Phenotype
MIM: 300846 MIM: 300171
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD99L2 PubMed Links

Gene Details

Gene
CD99L2
Gene Name
CD99 molecule like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184808.1 3567 UTR 3 NP_001171737.1
NM_001242614.1 3567 UTR 3 NP_001229543.1
NM_031462.3 3567 UTR 3 NP_113650.2
NM_134445.3 3567 UTR 3 NP_604394.1
NM_134446.3 3567 UTR 3 NP_604395.1
XM_011531202.2 3567 Intron XP_011529504.1
XM_017029890.1 3567 Intron XP_016885379.1
XM_017029891.1 3567 Intron XP_016885380.1
Gene
MTMR1
Gene Name
myotubularin related protein 1
There are no transcripts associated with this gene.

View Full Product Details