Product Details

SNP ID
rs3182038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69325020 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGAGCACAGAAAATACAGAAGA[G/T]GAAAAATACTTAGGATACAATACTA
Phenotype
MIM: 138292
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GFPT1 PubMed Links

Gene Details

Gene
GFPT1
Gene Name
glutamine--fructose-6-phosphate transaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244710.1 3354 UTR 3 NP_001231639.1
NM_002056.3 3354 UTR 3 NP_002047.2
XM_017003801.1 3354 UTR 3 XP_016859290.1
XM_017003802.1 3354 UTR 3 XP_016859291.1

View Full Product Details