Product Details

SNP ID
rs3192111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219289730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGAGAGGTTGAGCAGCGCATGG[G/T]GGGAGGGTGGTGGGAGGAGGCTGGG
Phenotype
MIM: 604139 MIM: 601773
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJB2 PubMed Links

Gene Details

Gene
DNAJB2
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
There are no transcripts associated with this gene.

Gene
MIR153-1
Gene Name
microRNA 153-1
There are no transcripts associated with this gene.

Gene
PTPRN
Gene Name
protein tyrosine phosphatase, receptor type N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199763.1 3220 UTR 3 NP_001186692.1
NM_001199764.1 3220 UTR 3 NP_001186693.1
NM_002846.3 3220 UTR 3 NP_002837.1
XM_011511566.1 3220 UTR 3 XP_011509868.1
XM_017004609.1 3220 UTR 3 XP_016860098.1
XM_017004610.1 3220 UTR 3 XP_016860099.1
XM_017004611.1 3220 UTR 3 XP_016860100.1
XM_017004612.1 3220 UTR 3 XP_016860101.1

View Full Product Details