Product Details

SNP ID
rs3195955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:114322789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTAAATGGAGTTTTTCTGACTAAC[A/G]AGGCTGACAACTTTCTGAACAATAT
Phenotype
MIM: 606025
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR568 PubMed Links

Gene Details

Gene
MIR568
Gene Name
microRNA 568
There are no transcripts associated with this gene.

Gene
ZBTB20
Gene Name
zinc finger and BTB domain containing 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164342.2 18663 UTR 3 NP_001157814.1
NM_001164343.2 18663 UTR 3 NP_001157815.1
NM_001164344.2 18663 UTR 3 NP_001157816.1
NM_001164345.2 18663 UTR 3 NP_001157817.1
NM_001164346.2 18663 UTR 3 NP_001157818.1
NM_001164347.2 18663 UTR 3 NP_001157819.1
NM_015642.5 18663 UTR 3 NP_056457.3

View Full Product Details