Product Details

SNP ID
rs3198762
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:649697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGCTCAACAACCCAGGAAAGCTG[G/T]GACGATTTCTTCAAATTGTTAAGAA
Phenotype
MIM: 616990 MIM: 188350
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLUL1 PubMed Links

Gene Details

Gene
CLUL1
Gene Name
clusterin like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289036.1 767 Intron NP_001275965.1
NM_001318522.1 767 Intron NP_001305451.1
NM_014410.4 767 Intron NP_055225.1
NM_199167.1 767 Intron NP_954636.1
XM_005258103.3 767 Intron XP_005258160.1
XM_006722322.2 767 Intron XP_006722385.1
XM_011525648.2 767 Intron XP_011523950.1
XM_011525649.1 767 Intron XP_011523951.1
XM_011525651.2 767 Intron XP_011523953.1
XM_011525652.1 767 Intron XP_011523954.1
XM_011525653.2 767 Intron XP_011523955.1
XM_017025707.1 767 Intron XP_016881196.1
Gene
TYMS
Gene Name
thymidylate synthetase
There are no transcripts associated with this gene.

Gene
TYMSOS
Gene Name
TYMS opposite strand
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012716.2 767 UTR 3 NP_001012734.2

View Full Product Details