Product Details

SNP ID
rs3138287
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:91146655 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCGTAAGCCACCACGACAGGCCC[A/C]ACTTTGAATAACTGACTACTTCTCA
Phenotype
MIM: 125255
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DCN PubMed Links

Gene Details

Gene
DCN
Gene Name
decorin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001920.4 Intron NP_001911.1
NM_133503.3 Intron NP_598010.1
NM_133504.3 Intron NP_598011.1
NM_133505.3 Intron NP_598012.1
NM_133506.3 Intron NP_598013.1
NM_133507.3 Intron NP_598014.1
XM_005268693.1 Intron XP_005268750.1
XM_006719270.1 Intron XP_006719333.1
XM_017018917.1 Intron XP_016874406.1

View Full Product Details