Product Details

SNP ID
rs3177209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7240429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCAAAATTCAGTTCAGCTTGG[G/T]GGTTTCCTCTTTATTGATGTGCCTC
Phenotype
MIM: 610684 MIM: 602151 MIM: 605125 MIM: 612910
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CTDNEP1 PubMed Links

Gene Details

Gene
CTDNEP1
Gene Name
CTD nuclear envelope phosphatase 1
There are no transcripts associated with this gene.

Gene
DVL2
Gene Name
dishevelled segment polarity protein 2
There are no transcripts associated with this gene.

Gene
GABARAP
Gene Name
GABA type A receptor-associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007278.1 Intron NP_009209.1
Gene
PHF23
Gene Name
PHD finger protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284517.1 Intron NP_001271446.1
NM_001284518.1 Intron NP_001271447.1
NM_024297.2 Intron NP_077273.2

View Full Product Details