Product Details

SNP ID
rs3181393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24315408 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTCACTGTATATTTTGCAATTG[A/G]GTCTGTGGACAGGAAGGGGCCGCAT
Phenotype
MIM: 600292 MIM: 604441 MIM: 601531 MIM: 605773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADCY4 PubMed Links

Gene Details

Gene
ADCY4
Gene Name
adenylate cyclase 4
There are no transcripts associated with this gene.

Gene
CIDEB
Gene Name
cell death-inducing DFFA-like effector b
There are no transcripts associated with this gene.

Gene
LTB4R
Gene Name
leukotriene B4 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143919.2 Intron NP_001137391.1
NM_181657.3 Intron NP_858043.1
Gene
LTB4R2
Gene Name
leukotriene B4 receptor 2
There are no transcripts associated with this gene.

Gene
NOP9
Gene Name
NOP9 nucleolar protein
There are no transcripts associated with this gene.

View Full Product Details