Product Details

SNP ID
rs3205785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67111552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTTGGAGAAACCAGGTGGTAC[A/T]GATTGCTGGTAAGGCCCTGCTGATG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C16orf70 PubMed Links

Gene Details

Gene
C16orf70
Gene Name
chromosome 16 open reading frame 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320540.1 348 Intron NP_001307469.1
NM_001320541.1 348 Intron NP_001307470.1
NM_001320542.1 348 Intron NP_001307471.1
NM_001320543.1 348 Intron NP_001307472.1
NM_025187.4 348 Intron NP_079463.2
XM_006721284.3 348 Intron XP_006721347.1
XM_011523350.2 348 Missense Mutation CAG,CTG Q14L XP_011521652.1
XM_017023729.1 348 Intron XP_016879218.1
XM_017023730.1 348 Intron XP_016879219.1
XM_017023731.1 348 Intron XP_016879220.1

View Full Product Details