Product Details

SNP ID
rs3207984
Assay Type
Functionally Tested
NCBI dbSNP Submissions
12
Location
Chr.6:32641437 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAGAGGAAGGAGACTGCCTGGCG[G/T]TGGCCTGAGTTCAGCAAATTTGGAG
Phenotype
MIM: 146880
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
HLA-DQA1 PubMed Links

Gene Details

Gene
HLA-DQA1
Gene Name
major histocompatibility complex, class II, DQ alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002122.3 266 Silent Mutation CGG,CGT R70R NP_002113.2
XM_006715079.3 266 Silent Mutation CGG,CGT R70R XP_006715142.1

View Full Product Details