Product Details

SNP ID
rs3732175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61178150 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCAGCCCTCCGCCGCAGAACCTC[C/G]GGAAGGGATGTCCTCTGCCCTGCGC
Phenotype
MIM: 615295
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AHSA2 PubMed Links

Gene Details

Gene
AHSA2
Gene Name
AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321300.1 733 UTR 5 NP_001308229.1
NM_001321301.1 733 UTR 5 NP_001308230.1
NM_001321302.1 733 Intron NP_001308231.1
NM_001321303.1 733 UTR 5 NP_001308232.1
NM_001321304.1 733 UTR 5 NP_001308233.1
NM_001321305.1 733 UTR 5 NP_001308234.1
NM_152392.4 733 UTR 5 NP_689605.1
XM_011532541.2 733 Intron XP_011530843.1
XM_011532543.2 733 UTR 5 XP_011530845.1
XM_011532545.2 733 Intron XP_011530847.1
XM_017003377.1 733 Intron XP_016858866.1
XM_017003378.1 733 Intron XP_016858867.1
Gene
USP34
Gene Name
ubiquitin specific peptidase 34
There are no transcripts associated with this gene.

View Full Product Details