Product Details

SNP ID
rs3209959
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:29148649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGGATTCAGACTTGGCATGTT[C/T]CCGCTCCTTTGACACGGAGCGGGAT
Phenotype
MIM: 601940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SRSF4 PubMed Links

Gene Details

Gene
SRSF4
Gene Name
serine and arginine rich splicing factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005626.4 1329 Missense Mutation AAA,GAA K416E NP_005617.2
XM_011541951.2 1329 Missense Mutation AAA,GAA K386E XP_011540253.1

View Full Product Details