Product Details

SNP ID
rs3733460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:146175842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCACCGCTGCGCGGGCCGACGAC[A/G]GGGCCGGGCGCAGCCCCAGGTTCGG
Phenotype
MIM: 607286
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LSM6 PubMed Links

Gene Details

Gene
LSM6
Gene Name
LSM6 homolog, U6 small nuclear RNA and mRNA degradation associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007080.2 Intron NP_009011.1
XM_017007672.1 Intron XP_016863161.1
XM_017007673.1 Intron XP_016863162.1

View Full Product Details