Product Details

SNP ID
rs3741737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52171538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGAGGCAGCTACAGGGAACATAA[G/T]GGGTAGGGGAGGGAAGGGGCTGGAG
Phenotype
MIM: 611161
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KRT80 PubMed Links

Gene Details

Gene
KRT80
Gene Name
keratin 80
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081492.1 1352 Silent Mutation CCA,CCC P418P NP_001074961.1
NM_182507.2 1352 Intron NP_872313.2
XM_005268676.3 1352 Intron XP_005268733.1

View Full Product Details