Product Details

SNP ID
rs3744398
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7251249 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAAGCTCCAGAGCTTGGCGGC[A/G]AAGGCCACGAACGTGCGCAGCCCCA
Phenotype
MIM: 609131 MIM: 610684 MIM: 615019 MIM: 605125
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CLDN7 PubMed Links

Gene Details

Gene
CLDN7
Gene Name
claudin 7
There are no transcripts associated with this gene.

Gene
CTDNEP1
Gene Name
CTD nuclear envelope phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143775.1 495 Intron NP_001137247.1
NM_015343.4 495 Silent Mutation TTC,TTT F16F NP_056158.2
Gene
ELP5
Gene Name
elongator acetyltransferase complex subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015362.4 495 Intron NP_056177.3
NM_203413.2 495 Intron NP_981958.1
NM_203414.2 495 Intron NP_981959.1
NM_203415.1 495 Intron NP_981960.1
XM_011523779.2 495 Intron XP_011522081.1
Gene
GABARAP
Gene Name
GABA type A receptor-associated protein
There are no transcripts associated with this gene.

View Full Product Details