Product Details

SNP ID
rs3213960
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38297491 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAGATACACAAAATGGCTTTCCT[A/T]AGCAGATACTTGCAGGCAAGAGTTC
Phenotype
MIM: 609368
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ATL2 PubMed Links

Gene Details

Gene
ATL2
Gene Name
atlastin GTPase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308076.1 Intron NP_001295005.1
XM_011533017.1 Intron XP_011531319.1
XM_011533018.2 Intron XP_011531320.1
XM_011533019.2 Intron XP_011531321.1
XM_011533020.1 Intron XP_011531322.1
XM_011533022.2 Intron XP_011531324.1
XM_011533023.2 Intron XP_011531325.1
XM_017004686.1 Intron XP_016860175.1
XM_017004687.1 Intron XP_016860176.1
XM_017004688.1 Intron XP_016860177.1

View Full Product Details